D-tert-Leucine (H-D-Tle-OH) ≥95%

Supplier: Matrix Scientific

Synonyms: H-D-Tle-OH

075411-1G 075411-5G 075411-10G
102541-228EA 355.38 USD
102541-228 102541-230 102541-232
D-tert-Leucine (H-D-Tle-OH) ≥95%
D-tert-Leucine (H-D-Tle-OH)

Hazard: Irritant

Formula: C₆H₁₃NO₂
MW: 131.2 g/mol
MDL Number: MFCD00004265
CAS Number: 26782-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR