(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol ≥95%

Supplier: Matrix Scientific

Synonyms: L-(+)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol, (1S,2S)-(+)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol

075410-10G 075410-100G
102541-224EA 64.62 USD
102541-224 102541-226
(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol ≥95%
(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol

Hazard: Irritant

Formula: C₉H₁₂N₂O₄
MW: 212.2026 g/mol
MDL Number: MFCD00007359
CAS Number: 2964-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR