Tyramine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxyphenethylamine, 4-(2-Aminoethyl)phenol

075409-10G 075409-100G
102541-220EA 364.62 USD
102541-220 102541-222
Tyramine ≥95%
Tyramine

Hazard: Irritant

Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 175…181 °C (8 mmHg)
Melting Pt: 160…162 °C
MDL Number: MFCD00008193
CAS Number: 51-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR