D-(-)-Ribose ≥95%
Supplier: Matrix Scientific
075407-100G
075407-500G
102541-214EA
93.85
USD
102541-214
102541-216
D-(-)-Ribose ≥95%
D(-)-Ribose
Hazard: Irritant
Formula:
C₅H₁₀O₅ MW: 150.13 g/mol |
MDL Number:
MFCD00135453 CAS Number: 50-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...