4-Amino-2-methyl-3-nitropyridine ≥95%

Supplier: Matrix Scientific

075404-250MG 075404-1G 075404-5G
102541-198EA 355.38 USD
102541-198 102541-200 102541-202
4-Amino-2-methyl-3-nitropyridine ≥95%
4-Amino-2-methyl-3-nitropyridine

Hazard: Irritant

Formula: C₆H₇N₃O₂
MW: 153.1400 g/mol
MDL Number: MFCD08277309
CAS Number: 27582-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR