2-Fluoro-5-nitrophenylboronic acid pinacol ester ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-5-nitrophenylboronic acid, pinacol ester, 2-(2-Fluoro-5-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Fluoro-5-nitrobenzeneboronic acid pinacol ester

075401-1G 075401-5G 075401-10G
102541-184EA 284.62 USD
102541-184 102541-186 102541-188
2-Fluoro-5-nitrophenylboronic acid pinacol ester ≥95%
2-Fluoro-5-nitrophenylboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₂H₁₅BFNO₄
MDL Number: MFCD06409955
CAS Number: 425378-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR