2,4,5,6-Tetraaminopyrimidine sulfate ≥95%
Supplier: Matrix Scientific
075399-100G
075399-25G
102541-178EA
116.92
USD
102541-178
102833-824
2,4,5,6-Tetraaminopyrimidine sulfate ≥95%
2,4,5,6-Tetraaminopyrimidine sulphate
Hazard: Irritant
Formula:
C₄H₈N₆·H₂SO₄ |
MDL Number:
MFCD00012786 CAS Number: 5392-28-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...