2-Chloro-3-nitrotoluene ≥95%

Supplier: Matrix Scientific

075394-5G 075394-25G
102541-154EA 93.85 USD
102541-154 102551-960
2-Chloro-3-nitrotoluene ≥95%
2-Chloro-3-nitrotoluene

Hazard: Irritant

Formula: C₇H₆ClNO₂
MDL Number: MFCD00134653
CAS Number: 3970-40-9
UN: 2433
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR