7-Nitro-1,2,3,4-tetrahydroisoquinoline ≥95%

Supplier: Matrix Scientific

Synonyms: 1,2,3,4-Tetrahydro-7-nitroisoquinoline

075392-1G 075392-5G 075392-10G
102541-140EA 116.92 USD
102541-140 102541-142 102541-144
7-Nitro-1,2,3,4-tetrahydroisoquinoline ≥95%
7-Nitro-1,2,3,4-tetrahydroisoquinoline

Hazard: Irritant

Formula: C₉H₁₀N₂O₂
MW: 178.1879 g/mol
MDL Number: MFCD04973400
CAS Number: 42923-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR