4,4'-Dimethyldiphenylamine ≥95%

Supplier: Matrix Scientific

Synonyms: p,p'-Ditolylamine, 4,4'-Dimethyldiphenylamine

075391-100G 075391-500G
102541-136EA 187.69 USD
102541-136 102541-138
4,4'-Dimethyldiphenylamine ≥95%
4,4'-Dimethyldiphenylamine

Hazard: Irritant

Formula: C₁₄H₁₅N
MW: 197.2756 g/mol
Melting Pt: 78…82 °C
MDL Number: MFCD00059317
CAS Number: 620-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR