2,2-Dimethylsuccinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2,2-Dimethyl-1,4-butanedioic acid, α,α-Dimethylsuccinic acid

075388-10G 075388-100G
102541-122EA 98.46 USD
102541-122 102541-124
2,2-Dimethylsuccinic acid ≥95%
2,2-Dimethylsuccinic acid

Hazard: Irritant

Formula: C₆H₁₀O₄
MW: 146.14 g/mol
MDL Number: MFCD00004196
CAS Number: 597-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR