3-Methyl-1H-pyrazole ≥95%
Supplier: Matrix Scientific
075384-100G
075384-25G
102541-106EA
126.15
USD
102541-106
102833-822
3-Methyl-1H-pyrazole ≥95%
3-Methyl-1H-pyrazole
Hazard: Irritant
Formula:
C₄H₆N₂ Boiling Pt: 203…205 °C Density: 1.02 Flash Pt: 93 °C |
MDL Number:
MFCD00005240 CAS Number: 1453-58-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...