(2-Bromophenyl)acetone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(2-Bromophenyl)propan-2-one

075383-1G 075383-5G
102541-100EA 290.77 USD
102541-100 102541-102
(2-Bromophenyl)acetone ≥95%
(2-Bromophenyl)acetone

Hazard: Irritant

Formula: C₉H₉BrO
MW: 213.07 g/mol
MDL Number: MFCD03410431
CAS Number: 21906-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR