trans-1,4-Cyclohexanedicarboxylic acid ≥95%
Supplier: Matrix Scientific
075381-10G
075381-100G
102541-090EA
23.08
USD
102541-090
102541-092
trans-1,4-Cyclohexanedicarboxylic acid ≥95%
trans-1,4-Cyclohexanedicarboxylic acid
Hazard: Irritant
Formula:
C₈H₁₂O₄ MW: 172.18 g/mol |
MDL Number:
MFCD00066203 CAS Number: 619-82-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...