Monomethyl succinate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methoxy-4-oxobutanoic acid, Methyl hydrogen succinate, Succinic acid monomethyl ester

075380-10G 075380-100G
102541-086EA 107.69 USD
102541-086 102541-088
Monomethyl succinate ≥95%
Monomethyl succinate

Hazard: Irritant

Formula: C₅H₈O₄
MW: 132.11 g/mol
MDL Number: MFCD00002788
CAS Number: 3878-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR