Calcium(2E,4E)hexa-2,4-dienoate ≥95%

Supplier: Matrix Scientific

Synonyms: Calcium Sorbate, Sorbic acid calcium salt, Calcium (2E,4E)-hexa-2,4-dienoate

075378-10G 075378-100G
102541-080EA 290.77 USD
102541-080 102541-082
Calcium(2E,4E)hexa-2,4-dienoate ≥95%
Calcium(2E,4E)hexa-2,4-dienoate

Hazard: Irritant

Formula: C₁₂H₁₄CaO₄
MW: 262.3200 g/mol
MDL Number: MFCD00043805
CAS Number: 7492-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR