4-Ethylcyclohexanone ≥95%

Supplier: Matrix Scientific

075377-10G 075377-100G
102541-076EA 72.31 USD
102541-076 102541-078
4-Ethylcyclohexanone ≥95%
4-Ethylcyclohexanone

Hazard: Irritant

Formula: C₈H₁₄O
MDL Number: MFCD00001644
CAS Number: 5441-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR