4-Amino-4H-1,2,4-triazole ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-1,2,4-triazole, 1,2,4-Triazol-4-ylamine, 4H-1,2,4-Triazol-4-amine

075373-100G 075373-25G
102541-062EA 72.31 USD
102541-062 102833-820
4-Amino-4H-1,2,4-triazole ≥95%
4-Amino-4H-1,2,4-triazole

Hazard: Irritant

Formula: C₂H₄N₄
MW: 84.08 g/mol
Melting Pt: 84…86 °C
MDL Number: MFCD00003099
CAS Number: 584-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR