2-Bromo-p-xylene ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-1,4-dimethylbenzene, 2,5-Dimethylbromobenzene

075371-10G 075371-100G
102541-052EA 47.69 USD
102541-052 102541-054
2-Bromo-p-xylene ≥95%
2-Bromo-p-xylene

Hazard: Irritant

Formula: C₈H₉Br
MW: 185.06 g/mol
MDL Number: MFCD00000074
CAS Number: 553-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR