2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl ≥95%

Supplier: Matrix Scientific

Synonyms: XPhos, 2-(Dicyclohexylphosphino)-2',4',6'-tri-i-propyl-1,1'-biphenyl, 2-Dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl

075369-5G 075369-25G
102541-042EA 73.85 USD
102541-042 102541-044
2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl ≥95%
2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl

Hazard: Irritant/Air Sensitive

Formula: C₃₃H₄₉P
MW: 476.72 g/mol
Melting Pt: 187…190 °C
MDL Number: MFCD04117682
CAS Number: 564483-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR