[5,5'-Biisobenzofuran]-1,1',3,3'-tetraone ≥95%
Supplier: Matrix Scientific
Synonyms:
5-(1,3-Dioxo-2-benzofuran-5-yl)-2-benzofuran-1,3-dione, 3,3',4,4'-Biphenyltetracarbocylic dianhydride. 4,4'-Biphthalic anhydride
075368-100G
075368-500G
102541-036EA
73.85
USD
102541-036
102541-038
[5,5'-Biisobenzofuran]-1,1',3,3'-tetraone ≥95%
[5,5'-Biisobenzofuran]-1,1',3,3'-tetraone
Hazard: Irritant
Formula:
C₁₆H₆O₆ Melting Pt: 299…305 °C |
MDL Number:
MFCD00039140 CAS Number: 2420-87-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...