2-Aminobenzophenone ≥95%

Supplier: Matrix Scientific

Synonyms: o-Aminobenzophenone

075365-10G 075365-100G
102541-022EA 64.62 USD
102541-022 102541-024
2-Aminobenzophenone ≥95%
2-Aminobenzophenone

Hazard: Irritant

Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
MDL Number: MFCD00007713
CAS Number: 2835-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR