(S)-Benzyl(2-oxotetrahydrofuran-3-yl)carbamate ≥95%

Supplier: Matrix Scientific

075357-1G 075357-5G
102540-984EA 43.08 USD
102540-984 102540-986
(S)-Benzyl(2-oxotetrahydrofuran-3-yl)carbamate ≥95%
(S)-Benzyl(2-oxotetrahydrofuran-3-yl)carbamate

Hazard: Irritant

Formula: C₁₂H₁₃NO₄
MW: 235.2400 g/mol
MDL Number: MFCD00674555
CAS Number: 35677-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR