3-Bromobenzene-1,2-diamine ≥95%

Supplier: Matrix Scientific

075355-1G 075355-5G
102540-974EA 166.15 USD
102540-974 102540-976
3-Bromobenzene-1,2-diamine ≥95%
3-Bromobenzene-1,2-diamine

Hazard: Irritant

Formula: C6H7BrN2
MDL Number: MFCD09842434
CAS Number: 1575-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR