Methyl-4-bromo-3-hydroxybenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-bromo-3-hydroxybenzoate

075354-1G 075354-5G
102540-970EA 76.92 USD
102540-970 102540-972
Methyl-4-bromo-3-hydroxybenzoate ≥95%
Methyl 4-bromo-3-hydroxybenzoate

Hazard: Irritant

Formula: C₈H₇BrO₃
MW: 231.0400 g/mol
MDL Number: MFCD08056372
CAS Number: 106291-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR