N-(Trimethylsilylmethyl)benzylamine ≥95%

Supplier: Matrix Scientific

075352-100G 075352-25G
102540-964EA 243.08 USD
102540-964 102833-818
N-(Trimethylsilylmethyl)benzylamine ≥95%
N-(Trimethylsilylmethyl)benzylamine

Hazard: Irritant

Formula: C₁₁H₁₉NSi
Boiling Pt: 89…90 °C (5 mmHg)
Density: 0.88
MDL Number: MFCD00012192
CAS Number: 53215-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR