tert-Butyl-1H-pyrazole-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 1H-Pyrazole, N1-BOC protected, tert-Butyl 1H-pyrazole-1-carboxylate

075351-1G 075351-5G
102540-954EA 32.31 USD
102540-954 102540-956
tert-Butyl-1H-pyrazole-1-carboxylate ≥95%
tert-Butyl-1H-pyrazole-1-carboxylate

Hazard: Irritant

Formula: C₈H₁₂N₂O₂
MW: 168.1900 g/mol
MDL Number: MFCD00800729
CAS Number: 219580-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR