ω,2,4-Trichloroacetophenone ≥95%

Supplier: Matrix Scientific

Synonyms: 2,4-Dichlorophenacyl chloride, α,2,4-Trichloroacetophenone, 2,2',4'-Trichloroacetophenone

075349-10G 075349-100G
102540-946EA 44.62 USD
102540-946 102540-948
ω,2,4-Trichloroacetophenone ≥95%
ω,2,4-Trichloroacetophenone

Hazard: Irritant

Formula: C₈H₅Cl₃O
MW: 223.49 g/mol
MDL Number: MFCD00000934
CAS Number: 4252-78-2
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR