2-(4-(tert-Butyl)phenyl)acetonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-tert-Butylphenylacetonitrile, 4-tert-Butylbenzyl cyanide

075343-100G 075343-25G
102540-926EA 104.62 USD
102540-926 102551-956
2-(4-(tert-Butyl)phenyl)acetonitrile ≥95%
2-(4-(tert-Butyl)phenyl)acetonitrile

Hazard: Irritant

Formula: C₁₂H₁₅N
MW: 173.2500 g/mol
Boiling Pt: 141-142°C
MDL Number: MFCD00128112
CAS Number: 3288-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR