D-(+)-Proline ≥95%

Supplier: Matrix Scientific

Synonyms: H-D-Pro-OH, (R)-(+)-Proline, D-Proline

075342-100G 075342-25G
102540-922EA 87.69 USD
102540-922 102833-814
D-(+)-Proline ≥95%
D-(+)-Proline

Hazard: Irritant/Hygroscopic

Formula: C₅H₉NO₂
MW: 115.13 g/mol
Melting Pt: 213…215 °C (dec.)
MDL Number: MFCD00064317
CAS Number: 344-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR