3,4-Dimethyldiphenylamine ≥95%

Supplier: Matrix Scientific

Synonyms: Phenyl(3,4-xylyl)amine, N-Phenyl-3,4-xylidine

075340-10G 075340-100G
102540-910EA 90.77 USD
102540-910 102540-912
3,4-Dimethyldiphenylamine ≥95%
3,4-Dimethyldiphenylamine

Hazard: Irritant

Formula: C14H15N
MW: 197.2800 g/mol
MDL Number: MFCD03093248
CAS Number: 17802-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR