2-Chloro-4,6-dimethylpyridine ≥95%

Supplier: Matrix Scientific

075339-5G 075339-25G
102540-906EA 61.54 USD
102540-906 102833-812
2-Chloro-4,6-dimethylpyridine ≥95%
2-Chloro-4,6-dimethylpyridine

Hazard: Irritant

Formula: C₇H₈ClN
MDL Number: MFCD08277279
CAS Number: 30838-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR