4-Methylbenzylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Aminomethyl)toluene

075337-10G 075337-100G
102540-896EA 55.38 USD
102540-896 102540-898
4-Methylbenzylamine ≥95%
4-Methylbenzylamine

Hazard: Corrosive/Air Sensitive

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 196-197°C
Melting Pt: 12-13°C
Density: 0.955
Flash Pt: 75°C
MDL Number: MFCD00008123
CAS Number: 104-84-7
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR