Tetrachlorophthalonitrile ≥95%

Supplier: Matrix Scientific

075334-1G 075334-5G 075334-10G
102540-882EA 72.31 USD
102540-882 102540-884 102540-886
Tetrachlorophthalonitrile ≥95%
Tetrachlorophthalonitrile

Hazard: Irritant

Formula: C₈Cl₄N₂
MW: 265.91 g/mol
MDL Number: MFCD00019740
CAS Number: 1953-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR