6-Bromo-2-naphthol ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-2-hydroxynaphthalene, 2-Bromo-6-hydroxynaphthalene

075333-100G 075333-500G
102540-880EA 75.38 USD
102540-880 102551-954
6-Bromo-2-naphthol ≥95%
6-Bromo-2-naphthol

Hazard: Irritant

Formula: C₁₀H₇BrO
MW: 223.07 g/mol
Melting Pt: 126-129°C
MDL Number: MFCD00004081
CAS Number: 15231-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR