4-Chloro-3-iodo-1-(4-methoxybenzyl)-1H-pyrazolo[4,3-c]pyridine ≥95%

Supplier: Matrix Scientific

075330-1G 075330-5G
102540-866EA 640 USD
102540-866 102540-868
4-Chloro-3-iodo-1-(4-methoxybenzyl)-1H-pyrazolo[4,3-c]pyridine ≥95%
4-Chloro-3-iodo-1-(4-methoxybenzyl)-1H-pyrazolo[4,3-c]pyridine

Hazard: Irritant

Formula: C₁₄H₁₁ClIN₃O
MW: 399.61 g/mol
MDL Number: MFCD19689569
CAS Number: 1246349-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR