tert-Butyl (3-hydroxycyclohexyl)carbamate ≥95%

Supplier: Matrix Scientific

075327-1G 075327-5G
102540-856EA 640 USD
102540-856 102540-858
tert-Butyl (3-hydroxycyclohexyl)carbamate ≥95%
tert-Butyl (3-hydroxycyclohexyl)carbamate

Hazard: Irritant

Formula: C₁₁H₂₁NO₃
MW: 215.2893 g/mol
MDL Number: MFCD06658389
CAS Number: 610302-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR