1,4-Diaminobutane dihydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
1,4-Butanediamine dihydrochloride, Tetramethylenediammonium dichloride, Tetramethylendiamine dihydrochloride, Putrescine dihydrochloride
075325-10G
075325-100G
102540-850EA
55.38
USD
102540-850
102540-852
1,4-Diaminobutane dihydrochloride ≥95%
1,4-Diaminobutane dihydrochloride
Hazard: Irritant
Formula:
C₄H₁₄Cl₂N₂ MW: 161.07 g/mol |
MDL Number:
MFCD00012526 CAS Number: 333-93-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...