3-Hydroxy-1,2,3-benzotriazin-4(3H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 3,4-Dihydro-3-hydroxy-4-oxo-1,2,3-benzotriazine, 3-Hydroxy-1,2,3-benzotriazin-4(3H)-one, HOOBt, 3-Hydroxy-4-ketobenzotriazine

075323-10G 075323-100G
102540-840EA 107.69 USD
102540-840 102540-842
3-Hydroxy-1,2,3-benzotriazin-4(3H)-one ≥95%
3-Hydroxy-1,2,3-benzotriazin-4(3H)-one

Hazard: Irritant

Formula: C7H5N3O2
MDL Number: MFCD00042803
CAS Number: 28230-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR