Methyl-5-methoxysalicylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-hydroxy-5-methoxybenzoate

075314-5G 075314-10G 075314-100G
102540-798EA 46.15 USD
102540-798 102540-800 102540-802
Methyl-5-methoxysalicylate ≥95%
Methyl 5-methoxysalicylate

Hazard: Irritant

Formula: C₉H₁₀O₄
MW: 182.1733 g/mol
MDL Number: MFCD00017185
CAS Number: 2905-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR