(R)-(-)-2-Amino-1-propanol ≥95%

Supplier: Matrix Scientific

Synonyms: D-Alaninol, D(-)-2-Amino-1-propanol, D(-)-Alaninol, (R)-2-Aminopropan-1-ol

075313-100G 075313-25G
102540-796EA 304.62 USD
102540-796 102833-806
(R)-(-)-2-Amino-1-propanol ≥95%
(R)-(-)-2-Amino-1-propanol

Hazard: Irritant

Formula: C₃H₉NO
MDL Number: MFCD00064413
CAS Number: 35320-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR