N-Boc-L-serine ≥95%

Supplier: Matrix Scientific

075311-100G 075311-25G
102540-788EA 100 USD
102540-788 102551-952
N-Boc-L-serine ≥95%
N-Boc-L-serine

Hazard: Irritant

Formula: C₈H₁₅NO₅
MDL Number: MFCD00037243
CAS Number: 3262-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR