Veratraldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 3,4-Dimethoxybenzaldehyde

075309-10G 075309-100G
102540-778EA 36.92 USD
102540-778 102540-780
Veratraldehyde ≥95%
Veratraldehyde

Hazard: Irritant/Air Sensitive

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 281 °C
Melting Pt: 42…46 °C
Flash Pt: 112 °C
MDL Number: MFCD00003363
CAS Number: 120-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR