(4-Methoxyphenyl)acetonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methoxybenzyl cyanide, 4-Methoxyphenylacetonitrile

075307-10G 075307-100G
102540-770EA 44.62 USD
102540-770 102540-772
(4-Methoxyphenyl)acetonitrile ≥95%
(4-Methoxyphenyl)acetonitrile

Hazard: Toxic

Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 285-287°C
Melting Pt: 8°C
Density: 1.085
Flash Pt: 117°C
MDL Number: MFCD00001919
CAS Number: 104-47-2
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR