Methyl-2-amino-5-(trifluoromethyl)benzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-amino-5-(trifluoromethyl)benzoate

075306-1G 075306-5G 075306-10G
102540-764EA 58.46 USD
102540-764 102540-766 102540-768
Methyl-2-amino-5-(trifluoromethyl)benzoate ≥95%
Methyl-2-amino-5-(trifluoromethyl)benzoate

Hazard: Irritant

Formula: C₉H₈F₃NO₂
MW: 219.16 g/mol
MDL Number: MFCD08234902
CAS Number: 117324-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR