L-Glutamic acid 5-methyl ester ≥95%
Supplier: Matrix Scientific
Synonyms:
H-Glu(OMe)-OH
075304-100G
075304-25G
102540-758EA
186.15
USD
102540-758
102833-804
L-Glutamic acid 5-methyl ester ≥95%
L-Glutamic acid 5-methyl ester
Hazard: Irritant
Formula:
C₆H₁₁NO₄ MW: 161.2 g/mol |
MDL Number:
MFCD00002632 CAS Number: 1499-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...