Methyl-2,3-diaminobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2,3-diaminobenzoate

075301-1G 075301-5G
102540-742EA 75.38 USD
102540-742 102540-744
Methyl-2,3-diaminobenzoate ≥95%
Methyl 2,3-diaminobenzoate

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MW: 166.1800 g/mol
MDL Number: MFCD04038589
CAS Number: 107582-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR