Ethyl-2-(triphenylphosphoranylidene)acetate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl triphenylphosphoranylidene-acetate, (Triphenylphosphoranylidene)acetic acid ethyl ester, (Carbethoxymethylene)triphenylphosphorane, Ethoxycarbonylmethylene-triphenylphosphorane, Ethyl 2-(triphenylphosphoranylidene)acetate

075298-100G 075298-500G
102540-730EA 121.54 USD
102540-730 102664-040
Ethyl-2-(triphenylphosphoranylidene)acetate ≥95%
Ethyl-2-(triphenylphosphoranylidene)acetate

Hazard: Irritant/Air Sensitive

Formula: C₂₂H₂₁O₂P
MW: 348.38 g/mol
Melting Pt: 126…130 °C
MDL Number: MFCD00009183
CAS Number: 1099-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR