1-Methylcyclopentanol ≥95%

Supplier: Matrix Scientific

075294-1G 075294-5G 075294-10G
102540-706EA 33.85 USD
102540-706 102540-708 102540-710
1-Methylcyclopentanol ≥95%
1-Methylcyclopentanol

Hazard: Irritant

Formula: C₆H₁₂O
MW: 100.16 g/mol
MDL Number: MFCD00001364
CAS Number: 1462-03-9
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR