L-(+)-Methyl-tert-leucinate hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
L-tert-Leucine methyl ester hydrochloride, H-Tle-OMe·HCl, H-Tle-OMe.HCl
075292-1G
075292-5G
102540-696EA
26.15
USD
102540-696
102540-698
L-(+)-Methyl-tert-leucinate hydrochloride ≥95%
L(+)-Methyl tert-leucinate hydrochloride
Hazard: Irritant
Formula:
C₇H₁₅NO₂·HCl Melting Pt: 183°C |
MDL Number:
MFCD00040604 CAS Number: 63038-27-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...